Order Entry
United Kingdom
ContactUsLinkComponent
2-(Allyloxy)-4-nitrobenzyl Bromide
2-(Allyloxy)-4-nitrobenzyl Bromide
Catalog # APOSOR470731-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Allyloxy)-4-nitrobenzyl Bromide
Catalog # APOSOR470731-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :272.095 g/mol
Storage Temperature:Ambient
CAS Number:1823359-23-4
Formula:C₁₀H₁₀BrNO₃
MDL Number:MFCD26937560