Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-(4-Pyridyl)propiolate
Methyl 3-(4-Pyridyl)propiolate
Methyl 3-(4-Pyridyl)propiolate
Methyl 3-(4-Pyridyl)propiolate
APOSOR470615-5G
:
Apollo Scientific
:
Methyl 3-(4-Pyridyl)propiolate
APOSOR470615-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
161.157 g/mol
:
Ambient
:
78584-33-5
:
C₉H₇NO₂
:
MFCD18205099