Ethyl 3-(2-Pyridyl)propiolate
Catalog # APOSOR470613-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:175.184 g/mol
Storage Temperature:Ambient
CAS Number:66869-70-3
Formula:C₁₀H₉NO₂
MDL Number:MFCD23381104