3-Hydroxy-4-methoxy-5-nitrobenzaldehyde
Catalog # APOSOR470435-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:197.145 g/mol
Storage Temperature:Ambient
CAS Number:80547-69-9
Formula:C₈H₇NO₅
MDL Number:MFCD02380161