Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Amino-3,4-dihydroquinolin-2(1H)-one
6-Amino-3,4-dihydroquinolin-2(1H)-one
6-Amino-3,4-dihydroquinolin-2(1H)-one
6-Amino-3,4-dihydroquinolin-2(1H)-one
APOSOR451145-5G
:
Apollo Scientific
:
6-Amino-3,4-dihydroquinolin-2(1H)-one
APOSOR451145-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
162.189 g/mol
:
Ambient
:
22246-13-5
:
C₉H₁₀N₂O
:
MFCD00559318