Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(5-Ethyl-1,3,4-oxadiazol-2-yl)benzaldehyde
4-(5-Ethyl-1,3,4-oxadiazol-2-yl)benzaldehyde
4-(5-Ethyl-1,3,4-oxadiazol-2-yl)benzaldehyde
4-(5-Ethyl-1,3,4-oxadiazol-2-yl)benzaldehyde
APOSOR42226-1G
:
Apollo Scientific
:
4-(5-Ethyl-1,3,4-oxadiazol-2-yl)benzaldehyde
APOSOR42226-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
202.209 g/mol
:
GHS07
:
Ambient
:
1309875-09-9
:
C₁₁H₁₀N₂O₂
:
MFCD19104824