Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-4-chlorophenylacetonitrile
2-Bromo-4-chlorophenylacetonitrile 97%
2-Bromo-4-chlorophenylacetonitrile 97%
2-Bromo-4-chlorophenylacetonitrile 97%
APOSOR400067-250MG
:
Apollo Scientific
:
2-Bromo-4-chlorophenylacetonitrile 97%
APOSOR400067-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
Environmentally Preferable:
:
2-(2-Bromo-4-chlorophenyl)acetonitrile
:
:
230.489 g/mol
:
Ambient
:
52864-54-7
:
C₈H₅BrClN
:
MFCD11044834