5-Bromo-2-nitrophenylacetonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:125914-22-9
:
Formula:C₈H₅BrN₂O₂
MDL Number:MFCD03206463
Molecular Weight:241.041 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1 III
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together