Order Entry
United Kingdom
ContactUsLinkComponent
1-((4R,5R)-2,2-dimethyl-5-vinyl-1,3-dioxolan-4-yl)ethanone ≥98%
1-((4R,5R)-2,2-dimethyl-5-vinyl-1,3-dioxolan-4-yl)ethanone ≥98%
Catalog # APOSOR370052-1G
Supplier:  Apollo Scientific
CAS Number:  
1-((4R,5R)-2,2-dimethyl-5-vinyl-1,3-dioxolan-4-yl)ethanone ≥98%
Catalog # APOSOR370052-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :170.206 g/mol
Storage Temperature:Ambient
CAS Number:1042237-51-3
Formula:C₉H₁₄O₃
MDL Number:MFCD19443200