Order Entry
United Kingdom
ContactUsLinkComponent
9-Bromo-7-methyl-2-morpholino-4H-pyrido[1,2-a]pyrimidin-4-one ≥98%
9-Bromo-7-methyl-2-morpholino-4H-pyrido[1,2-a]pyrimidin-4-one ≥98%
Catalog # APOSOR350434-100MG
Supplier:  Apollo Scientific
CAS Number:  
9-Bromo-7-methyl-2-morpholino-4H-pyrido[1,2-a]pyrimidin-4-one ≥98%
Catalog # APOSOR350434-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :324.173 g/mol
Storage Temperature:Ambient
CAS Number:351002-16-9
Formula:C₁₃H₁₄BrN₃O₂
MDL Number:MFCD11113210