Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(2-Chloropyridin-4-yl)piperazine
1-(2-Chloropyridin-4-yl)piperazine
1-(2-Chloropyridin-4-yl)piperazine
APOSOR350092-1G
:
Apollo Scientific
:
1-(2-Chloropyridin-4-yl)piperazine
APOSOR350092-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
197.665 g/mol
:
Ambient
:
854159-45-8
:
C₉H₁₂ClN₃
:
MFCD06738480