(4-Chloro-3-hydroxy-phenyl)-carbamic acid tert-butyl ester
Catalog # APOSOR346370-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:243.687 g/mol
Storage Temperature:Ambient
CAS Number:345893-27-8
Formula:C₁₁H₁₄ClNO₃
MDL Number:MFCD09951954