Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-N-phenyl-acetamide
2-Bromo-N-phenyl-acetamide
2-Bromo-N-phenyl-acetamide
2-Bromo-N-phenyl-acetamide
APOSOR346345-1G
:
Apollo Scientific
:
2-Bromo-N-phenyl-acetamide
APOSOR346345-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
Environmentally Preferable:
:
2-Bromo-N-phenylacetamide
:
:
214.059 g/mol
:
Ambient
:
5326-87-4
:
C₈H₈BrNO
:
MFCD00462518