Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-6-methoxyisonicotinic acid
2-Bromo-6-methoxyisonicotinic acid 97%
2-Bromo-6-methoxyisonicotinic acid 97%
2-Bromo-6-methoxyisonicotinic acid 97%
APOSOR345428-250MG
:
Apollo Scientific
:
2-Bromo-6-methoxyisonicotinic acid 97%
APOSOR345428-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
232.031 g/mol
:
Ambient
:
853029-93-3
:
C₇H₆BrNO₃
:
MFCD11101033