Ethyl 5-formyl-1-benzofuran-2-carboxylate, tech
Catalog # APOSOR33447-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:218.205 g/mol
Storage Temperature:Ambient
CAS Number:10035-37-7
Formula:C₁₂H₁₀O₄
MDL Number:MFCD11501717