Order Entry
United Kingdom
ContactUsLinkComponent
4-[(E)-2-(Dimethylamino)ethenyl]-2-oxo-2H-chromene-3-carbonitrile, tech
4-[(E)-2-(Dimethylamino)ethenyl]-2-oxo-2H-chromene-3-carbonitrile, tech
Catalog # APOSOR33260-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[(E)-2-(Dimethylamino)ethenyl]-2-oxo-2H-chromene-3-carbonitrile, tech
Catalog # APOSOR33260-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :240.257 g/mol
Storage Temperature:Ambient
CAS Number:209617-29-8
Formula:C₁₄H₁₂N₂O₂
MDL Number:MFCD01304441