Order Entry
United Kingdom
ContactUsLinkComponent
5-Chloro-2-oxo-N-phenyl-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carboxamide, tech
5-Chloro-2-oxo-N-phenyl-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carboxamide, tech
Catalog # APOSOR33220-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-2-oxo-N-phenyl-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carboxamide, tech
Catalog # APOSOR33220-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :286.713 g/mol
Storage Temperature:Ambient
CAS Number:339028-41-0
Formula:C₁₅H₁₁ClN₂O₂
MDL Number:MFCD00141340