6-[(2-Methylphenyl)methyl]pyridazin-3-ol, tech
Catalog # APOSOR33212-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:200.236 g/mol
Storage Temperature:Ambient
CAS Number:60932-69-6
Formula:C₁₂H₁₂N₂O
MDL Number:MFCD00214802