Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-[(3-Methylphenyl)methyl]pyridazin-3-ol
6-[(3-Methylphenyl)methyl]pyridazin-3-ol, tech
6-[(3-Methylphenyl)methyl]pyridazin-3-ol, tech
APOSOR33067-1G
:
Apollo Scientific
:
6-[(3-Methylphenyl)methyl]pyridazin-3-ol, tech
APOSOR33067-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
Environmentally Preferable:
:
:
200.236 g/mol
:
Ambient
:
339008-34-3
:
C₁₂H₁₂N₂O
:
MFCD00214779