Order Entry
United Kingdom
ContactUsLinkComponent
1-Phenyl-3-(pyridin-3-yl)-4,5-dihydro-1H-pyrazol-5-one, tech
1-Phenyl-3-(pyridin-3-yl)-4,5-dihydro-1H-pyrazol-5-one, tech
Catalog # APOSOR32843-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Phenyl-3-(pyridin-3-yl)-4,5-dihydro-1H-pyrazol-5-one, tech
Catalog # APOSOR32843-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :237.257 g/mol
Storage Temperature:Ambient
CAS Number:21683-61-4
Formula:C₁₄H₁₁N₃O
MDL Number:MFCD00665192