(Pyridin-3-yl)methyl 8-methoxy-2H-chromene-3-carboxylate, tech
Catalog # APOSOR32727-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:297.305 g/mol
Storage Temperature:Ambient
CAS Number:338419-73-1
Formula:C₁₇H₁₅NO₄
MDL Number:MFCD00202026