6-Chloro-2-(2-chloroethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one, tech
Catalog # APOSOR32684-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:246.09 g/mol
Storage Temperature:Ambient
CAS Number:865658-35-1
Formula:C₁₀H₉Cl₂NO₂
MDL Number:MFCD03787075