Order Entry
United Kingdom
ContactUsLinkComponent
 
4-(Benzenesulphonyl)-3-methyl-3,4-dihydro-2H-1,4-benzoxazine, tech
Catalog # APOSOR32641-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Benzenesulphonyl)-3-methyl-3,4-dihydro-2H-1,4-benzoxazine, tech
Catalog # APOSOR32641-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :289.35 g/mol
Storage Temperature:Ambient
CAS Number:478031-97-9
Formula:C₁₅H₁₅NO₃S
MDL Number:MFCD02083423