1-Cyclopropyl-6-oxo-2-phenylpiperidine-3-carboxylic acid, tech
Catalog # APOSOR32571-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:259.3 g/mol
Storage Temperature:Ambient
CAS Number:477890-39-4
Formula:C₁₅H₁₇NO₃
MDL Number:MFCD02571879