6-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridine-3-carbonitrile, tech
Catalog # APOSOR32510-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:197.236 g/mol
Storage Temperature:Ambient
CAS Number:861207-98-9
Formula:C₁₂H₁₁N₃
MDL Number:MFCD04124853