Order Entry
United Kingdom
ContactUsLinkComponent
(2R,3R)-3-((S)-1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoic acid
(2R,3R)-3-((S)-1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoic acid
Catalog # APOSOR323233-250MG
Supplier:  Apollo Scientific
CAS Number:  
(2R,3R)-3-((S)-1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoic acid
Catalog # APOSOR323233-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :287.352 g/mol
Storage Temperature:Ambient
CAS Number:120205-50-7
Formula:C₁₄H₂₅NO₅
MDL Number:MFCD18157735