Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-4-chlorobenzyl alcohol
3-Bromo-4-chlorobenzyl alcohol 97%
3-Bromo-4-chlorobenzyl alcohol 97%
APOSOR322285-1G
:
Apollo Scientific
:
3-Bromo-4-chlorobenzyl alcohol 97%
APOSOR322285-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
221.479 g/mol
:
100 °C (0.15 mm Hg)
:
GHS07
:
Ambient
:
329218-12-4
:
C₇H₆BrClO
:
MFCD08059085