Order Entry
United Kingdom
ContactUsLinkComponent
1-(6-Chloropyridin-2-yl)cyclobutanecarbonitrile 95%
1-(6-Chloropyridin-2-yl)cyclobutanecarbonitrile 95%
Catalog # APOSOR315880-100MG
Supplier:  Apollo Scientific
CAS Number:  
1-(6-Chloropyridin-2-yl)cyclobutanecarbonitrile 95%
Catalog # APOSOR315880-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :192.645 g/mol
Storage Temperature:Ambient
CAS Number:1190644-32-6
Formula:C₁₀H₉ClN₂
MDL Number:MFCD22581609