Order Entry
United Kingdom
ContactUsLinkComponent
3-Iodo-2-nitropyridine 98%
3-Iodo-2-nitropyridine 98%
Catalog # APOSOR315076-5G
Supplier:  Apollo Scientific
CAS Number:  
3-Iodo-2-nitropyridine 98%
Catalog # APOSOR315076-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :249.994 g/mol
Storage Temperature:Refrigerator
CAS Number:54231-34-4
Formula:C₅H₃IN₂O₂
MDL Number:MFCD08061318