£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
Ethyl 6-bromo-4-chloroquinazoline-2-carboxylate
Ethyl 6-bromo-4-chloroquinazoline-2-carboxylate
Ethyl 6-bromo-4-chloroquinazoline-2-carboxylate
APOSOR309396-100MG
:
Apollo Scientific
:
Ethyl 6-bromo-4-chloroquinazoline-2-carboxylate
APOSOR309396-100MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mg
Seq:
2440
:
:
315.55 g/mol
:
GHS07
:
Ambient
:
1159976-38-1
:
C₁₁H₈BrClN₂O₂
:
MFCD12026446