4-Hydroxy-2-methylquinoline-6-carboxylic acid ethyl ester
Catalog # APOSOR309246-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: Ethyl 4-hydroxy-2-methylquinoline-6-carboxylate
:
:231.247 g/mol
Storage Temperature:Ambient
CAS Number:300590-94-7
Formula:C₁₃H₁₃NO₃
MDL Number:MFCD00440392