tert-Butyl (3S)-3-(methylamino)pyrrolidine-1-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:147081-59-2
:
Formula:C₁₀H₂₀N₂O₂
MDL Number:MFCD09752962
:200.278 g/mol
Storage Temperature:Ambient
Frequently Bought Together