4-Hydroxy-8-methoxyquinoline
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:21269-34-1
- Formula:C₁₀H₉NO₂
- MDL Number:MFCD02684163
- Molecular Weight:175.184 g/mol
- Storage Temperature:Ambient
5 Options Available Below
Product Details & Documents
- CAS Number:21269-34-1
- Formula:C₁₀H₉NO₂
- MDL Number:MFCD02684163
- Molecular Weight:175.184 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



