8-Methyl-quinoline-3-carboxylic acid
Catalog # APOSOR307848-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:187.195 g/mol
Storage Temperature:Ambient
CAS Number:71082-55-8
Formula:C₁₁H₉NO₂
MDL Number:MFCD08437582