
trans-3-Benzylamino-4-hydroxy-pyrrolidine-1-carboxylic acid tert-butyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:138026-89-8
:
Formula:C₁₆H₂₄N₂O₃
MDL Number:MFCD09608005
:292.373 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together