trans-3-Methoxycyclobutane-1-methamine hydrochloride
Catalog # APOSOR304380-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:151.634 g/mol
Storage Temperature:Ambient
CAS Number:2094029-36-2
Formula:C₆H₁₃NO·ClH
MDL Number:MFCD30734068