2-Methoxyquinolin-6-amine
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:1153800-77-1
- Formula:C₁₀H₁₀N₂O
- MDL Number:MFCD12173233
- Molecular Weight:174.199 g/mol
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
- CAS Number:1153800-77-1
- Formula:C₁₀H₁₀N₂O
- MDL Number:MFCD12173233
- Molecular Weight:174.199 g/mol
- Storage Temperature:Ambient



