
1-(3-Chloropyridin-2-yl)ethanone
Catalog # APOSOR302156-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:155.582 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:131109-75-6
Formula:C₇H₆ClNO
MDL Number:MFCD11656222