
3H-Imidazo[4,5-c]pyridine 5-oxide
Catalog # APOSOR302028-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:135.123 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:91184-02-0
Formula:C₆H₅N₃O
MDL Number:MFCD22989413