Order Entry
United Kingdom
ContactUsLinkComponent
2-Methyl-5-iodopyridine
2-Methyl-5-iodopyridine
Catalog # APOSOR301057-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-5-iodopyridine
Catalog # APOSOR301057-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :219.023 g/mol
Storage Temperature:Ambient
CAS Number:695-17-0
Formula:C₆H₆IN
MDL Number:MFCD07437990