
1-(5-Bromo-3-chloropyridin-2-yl)ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1256808-29-3
:
Formula:C₇H₅BrClNO
MDL Number:MFCD18257815
:234.478 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together