
5-Phenyl-1,3,4-oxadiazole-2-carbonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:98591-60-7
:
Formula:C₉H₅ClN₂O₂
MDL Number:MFCD03086204
:208.601 g/mol
Melting Pt:80…90 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together