
5-Chloro-4-methoxythiophene-3-carbonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175137-49-2
:
Formula:C₆H₄Cl₂O₂S
MDL Number:MFCD00068000
:211.066 g/mol
:GHS05
Storage Temperature:Refrigerator
Frequently Bought Together