Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2,3-Trimethyldecane
2,2,3-Trimethyldecane
2,2,3-Trimethyldecane
APOSOR2376-1G
:
Apollo Scientific
:
2,2,3-Trimethyldecane
APOSOR2376-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
184.361 g/mol
:
Ambient
:
1993
:
3,III
:
GHS02
:
62338-09-4
:
C₁₃H₂₈
:
MFCD03788713