
5-Bromo-2-methyl-4-nitro-1H-imidazole
Catalog # APOSOR21890-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.997 g/mol
Melting Pt:256…257 °C
:GHS07
Storage Temperature:Ambient
CAS Number:18874-52-7
Formula:C₄H₄BrN₃O₂
MDL Number:MFCD00156130