Order Entry
United Kingdom
ContactUsLinkComponent
2-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridin-3-ol
Irritant
2-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridin-3-ol
Catalog # APOSOR21535-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridin-3-ol
Catalog # APOSOR21535-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :188.226 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:175135-48-5
Formula:C₁₁H₁₂N₂O
MDL Number:MFCD00067779