Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Benzo[b]furan-2-yl)-1H-pyrazole
3-(Benzo[b]furan-2-yl)-1H-pyrazole
3-(Benzo[b]furan-2-yl)-1H-pyrazole
3-(Benzo[b]furan-2-yl)-1H-pyrazole
APOSOR183462-1G
:
Apollo Scientific
:
3-(Benzo[b]furan-2-yl)-1H-pyrazole
APOSOR183462-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
Environmentally Preferable:
:
:
184.194 g/mol
:
Ambient
:
666728-39-8
:
C₁₁H₈N₂O
:
MFCD00112205