
2-Amino-5-hydroxy-1,3-benzothiazole
Catalog # APOSOR17933-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:166.2 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:118526-19-5
Formula:C₇H₆N₂OS
MDL Number:MFCD00985125