
8-Amino-2,4-dimethyl-2H-1,4-benzoxazin-3(4H)-one 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:870064-85-0
:
Formula:C₁₀H₁₂N₂O₂
MDL Number:MFCD09998375
:192.214 g/mol
Melting Pt:94 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together