
2-Bromo-1-(2-chloropyridin-4-yl)ethanone hydrobromide
Catalog # APOSOR111611-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:315.39 g/mol
Storage Temperature:Ambient
UN:2923
ADR:8+6.1,III
:GHS05
CAS Number:569667-89-6
Formula:C₇H₅BrClNO·BrH
MDL Number:MFCD13192035